Assay Details
| Gene Symbol: pak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002717.1 | NP_001002717.1 | 10 - 11 | 1149 | 77 | |
| XM_005171262.4 | XP_005171319.1 | 10 - 11 | 1472 | 77 | ||
| XM_005171263.4 | XP_005171320.1 | 11 - 12 | 1473 | 77 | ||
| GenBank mRNA | BC076511.1 | - | 10 - 11 | 1149 | 77 | |
| BC165251.1 | - | 9 - 10 | 1064 | 77 | ||
| DQ656110.1 | - | 9 - 10 | 979 | 77 | ||
More Information
Set Membership: |
|

pak2a
Genomic Map