Assay Details
| Gene Symbol: pak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002717.1 | NP_001002717.1 | 13 - 14 | 1583 | 63 | |
| XM_005171262.4 | XP_005171319.1 | 13 - 14 | 1906 | 63 | ||
| XM_005171263.4 | XP_005171320.1 | 14 - 15 | 1907 | 63 | ||
| GenBank mRNA | BC076511.1 | - | 13 - 14 | 1583 | 63 | |
| BC165251.1 | - | 12 - 13 | 1498 | 63 | ||
| DQ656110.1 | - | 12 - 13 | 1413 | 63 | ||
More Information
Set Membership: |
|

pak2a
Genomic Map