Assay Details
| Gene Symbol: pak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002717.1 | NP_001002717.1 | 3 - 4 | 413 | 84 | |
| XM_005171262.4 | XP_005171319.1 | 3 - 4 | 736 | 84 | ||
| XM_005171263.4 | XP_005171320.1 | 4 - 5 | 737 | 84 | ||
| GenBank mRNA | BC076511.1 | - | 3 - 4 | 413 | 84 | |
| BC165251.1 | - | 2 - 3 | 328 | 84 | ||
More Information
Set Membership: |
|

pak2a
Genomic Map