Assay Details
| Gene Symbol: pak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002717.1 | NP_001002717.1 | 5 - 6 | 667 | 98 | |
| XM_005171262.4 | XP_005171319.1 | 5 - 6 | 990 | 98 | ||
| XM_005171263.4 | XP_005171320.1 | 6 - 7 | 991 | 98 | ||
| GenBank mRNA | BC076511.1 | - | 5 - 6 | 667 | 98 | |
| BC165251.1 | - | 4 - 5 | 582 | 98 | ||
More Information
Set Membership: |
|

pak2a
Genomic Map