Assay Details
| Gene Symbol: pak2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002717.1 | NP_001002717.1 | 9 - 10 | 1032 | 73 | |
| XM_005171262.4 | XP_005171319.1 | 9 - 10 | 1355 | 73 | ||
| XM_005171263.4 | XP_005171320.1 | 10 - 11 | 1356 | 73 | ||
| GenBank mRNA | BC076511.1 | - | 9 - 10 | 1032 | 73 | |
| BC165251.1 | - | 8 - 9 | 947 | 73 | ||
| DQ656110.1 | - | 8 - 9 | 862 | 73 | ||
More Information
Set Membership: |
|

pak2a
Genomic Map