Assay Details
| Gene Symbol: dpy19l1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002868.1 | NP_001002868.1 | 2 - 3 | 351 | 158 | |
| GenBank mRNA | AY648728.1 | - | 2 - 3 | 351 | 158 | |
| BC163516.1 | - | 2 - 3 | 324 | 158 | ||
| BC163534.1 | - | 2 - 3 | 324 | 158 | ||
More Information
Set Membership: |
|

dpy19l1l
Genomic Map