Assay Details
| Gene Symbol: rpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003447.1 | NP_001003447.1 | 2 - 3 | 330 | 82 | |
| XM_021468049.1 | XP_021323724.1 | 2 - 3 | 354 | 82 | ||
| GenBank mRNA | BC075894.1 | - | 2 - 3 | 330 | 82 | |
| BC155095.1 | - | 1167 | 82 | |||
More Information
Set Membership: |
|

rpl15
Genomic Map