Assay Details
| Gene Symbol: dnal4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003603.1 | NP_001003603.1 | 2 - 3 | 159 | 75 | |
| GenBank mRNA | BC077142.1 | - | 2 - 3 | 159 | 75 | |
More Information
Set Membership: |
|

dnal4a
Genomic Map