Assay Details
| Gene Symbol: timm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003637.3 | NP_001003637.1 | 1 - 2 | 236 | 68 | |
| GenBank mRNA | BC077084.1 | - | 1 - 2 | 218 | 68 | |
More Information
Set Membership: |
|

timm8a
Genomic Map