Assay Details
| Gene Symbol: ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113650.1 | NP_001107122.1 | 1 - 2 | 212 | 129 | |
| GenBank mRNA | BC079510.1 | - | 1 - 2 | 202 | 129 | |
| BC157378.1 | - | 1 - 2 | 198 | 129 | ||
| CK688635.1 | - | 1 - 2 | 202 | 129 | ||
More Information
Set Membership: |
|

ndufaf2
Genomic Map