Assay Details
| Gene Symbol: mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003778.2 | NP_001003778.1 | 8 - 9 | 702 | 62 | |
| GenBank mRNA | BC079488.1 | - | 8 - 9 | 706 | 62 | |
More Information
Set Membership: |
|

mrps27
Genomic Map