Assay Details
| Gene Symbol: ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003826.1 | NP_001003826.1 | 1 - 2 | 141 | 75 | |
| XM_005158761.4 | XP_005158818.1 | 2 - 3 | 335 | 75 | ||
| GenBank mRNA | BC114244.1 | - | 2 - 3 | 267 | 75 | |
More Information
Set Membership: |
|

ap4s1
Genomic Map