Assay Details
| Gene Symbol: rpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003861.1 | NP_001003861.1 | 2 - 3 | 167 | 77 | |
| GenBank mRNA | AY141976.1 | - | 2 - 3 | 146 | 77 | |
| AY648736.1 | - | 2 - 3 | 167 | 77 | ||
| BC090911.1 | - | 3 - 4 | 251 | 77 | ||
| BC115139.1 | - | 3 - 4 | 193 | 77 | ||
| BC152564.1 | - | 3 - 4 | 176 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC152564.1 transcripts.
Set Membership: |
|

rpl9
Genomic Map