Assay Details
| Gene Symbol: fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004506.1 | NP_001004506.1 | 3 - 4 | 491 | 84 | |
| GenBank mRNA | BC163638.1 | - | 3 - 4 | 577 | 84 | |
| BC163644.1 | - | 3 - 4 | 577 | 84 | ||
| BC163849.1 | - | 3 - 4 | 577 | 84 | ||
More Information
Set Membership: |
|

fkbp7
Genomic Map