Assay Details
| Gene Symbol: ppp1cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004527.3 | NP_001004527.2 | 5 - 6 | 611 | 64 | |
| GenBank mRNA | BC095825.1 | - | 5 - 6 | 609 | 64 | |
| BC116539.1 | - | 5 - 6 | 611 | 64 | ||
More Information
Set Membership: |
|

ppp1cb
Genomic Map