Assay Details
| Gene Symbol: pon1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004542.1 | NP_001004542.1 | 6 - 7 | 751 | 97 | |
| GenBank mRNA | BC080844.1 | - | 6 - 7 | 751 | 97 | |
| BC164584.1 | - | 6 - 7 | 728 | 97 | ||
More Information
Set Membership: |
|

pon1
Genomic Map