Assay Details
| Gene Symbol: mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005949.1 | NP_001005949.1 | 6 - 7 | 766 | 84 | |
| GenBank mRNA | BC083455.1 | - | 6 - 7 | 766 | 84 | |
More Information
Set Membership: |
|

mettl5
Genomic Map