Assay Details
| Gene Symbol: dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007056.1 | NP_001007057.1 | 1 - 2 | 137 | 63 | |
| GenBank mRNA | BC083277.1 | - | 1 - 2 | 137 | 63 | |
| BC164599.1 | - | 1 - 2 | 129 | 63 | ||
More Information
Set Membership: |
|

dck
Genomic Map