Assay Details
| Gene Symbol: oxct1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007291.1 | NP_001007292.1 | 3 - 4 | 305 | 91 | |
| GenBank mRNA | BC078408.2 | - | 3 - 4 | 305 | 91 | |
| BC163972.1 | - | 3 - 4 | 332 | 91 | ||
More Information
Set Membership: |
|

oxct1a
Genomic Map