Assay Details
| Gene Symbol: neil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007335.1 | NP_001007336.1 | 3 - 4 | 449 | 73 | |
| XM_005157318.3 | XP_005157375.1 | 3 - 4 | 508 | 73 | ||
| GenBank mRNA | BC085550.1 | - | 3 - 4 | 449 | 73 | |
| BC155157.1 | - | 3 - 4 | 429 | 73 | ||
| BC164271.1 | - | 3 - 4 | 381 | 73 | ||
More Information
Set Membership: |
|

neil3
Genomic Map