Assay Details
| Gene Symbol: snrpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007383.2 | NP_001007384.2 | 2 - 3 | 367 | 60 | |
| GenBank mRNA | BC085450.1 | - | 2 - 3 | 352 | 60 | |
| BC165478.1 | - | 2 - 3 | 264 | 60 | ||
| EH529176.1 | - | 7 - 8 | 735 | 60 | ||
| EH548169.1 | - | 2 - 3 | 391 | 60 | ||
More Information
Set Membership: |
|

snrpa1
Genomic Map