Assay Details
| Gene Symbol: snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008581.1 | NP_001008581.1 | 2 - 3 | 724 | 86 | |
| XM_009297590.3 | XP_009295865.1 | 2 - 3 | 720 | 86 | ||
| GenBank mRNA | BC086969.1 | - | 2 - 3 | 724 | 86 | |
More Information
Set Membership: |
|

snai2
Genomic Map