Assay Details
| Gene Symbol: fgf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009564.2 | NP_001009564.2 | 1 - 2 | 256 | 70 | |
| GenBank mRNA | AY753222.1 | - | 1 - 2 | 256 | 70 | |
| BC162690.1 | - | 1 - 2 | 315 | 70 | ||
| BC162711.1 | - | 1 - 2 | 315 | 70 | ||
More Information
Set Membership: |
|

fgf23
Genomic Map