Assay Details
| Gene Symbol: rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012250.2 | NP_001012250.1 | 1 - 2 | 334 | 108 | |
| GenBank mRNA | AY314756.1 | - | 1 - 2 | 256 | 108 | |
| BC098604.1 | - | 1 - 2 | 333 | 108 | ||
| BC154349.1 | - | 1 - 2 | 334 | 108 | ||
| BC171441.1 | - | 1 - 2 | 323 | 108 | ||
| BC171443.1 | - | 1 - 2 | 323 | 108 | ||
More Information
Set Membership: |
|

rhov
Genomic Map