Assay Details
| Gene Symbol: fut10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012374.1 | NP_001012374.1 | 2 - 3 | 335 | 71 | |
| XM_005155462.4 | XP_005155519.1 | 2 - 3 | 595 | 71 | ||
| XM_009297433.3 | XP_009295708.1 | 3 - 4 | 663 | 71 | ||
| GenBank mRNA | AJ879586.1 | - | 2 - 3 | 335 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009297433.3 transcripts.
Set Membership: |
|

fut10
Genomic Map