Assay Details
| Gene Symbol: mtrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012494.1 | NP_001012512.1 | 5 - 6 | 738 | 55 | |
| GenBank mRNA | BC090173.1 | - | 5 - 6 | 738 | 55 | |
More Information
Set Membership: |
|

mtrf1
Genomic Map