Assay Details
| Gene Symbol: tnfsf10l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013283.1 | NP_001013301.1 | 2 - 3 | 455 | 112 | |
| XM_005165144.3 | XP_005165201.1 | 2 - 3 | 461 | 112 | ||
| GenBank mRNA | BC090312.1 | - | 2 - 3 | 455 | 112 | |
| DQ812114.1 | - | 2 - 3 | 303 | 112 | ||
More Information
Set Membership: |
|

tnfsf10l4
Genomic Map