Assay Details
| Gene Symbol: znf45l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013496.1 | NP_001013514.1 | 2 - 3 | 380 | 105 | |
| XM_017353122.2 | XP_017208611.1 | 3 - 4 | 1346 | 105 | ||
| XM_017353123.2 | XP_017208612.1 | 2 - 3 | 1150 | 105 | ||
| GenBank mRNA | BC090439.1 | - | 2 - 3 | 380 | 105 | |
More Information
Set Membership: |
|

znf45l
Genomic Map