Assay Details
| Gene Symbol: enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013527.1 | NP_001013545.1 | 5 - 6 | 977 | 61 | |
| GenBank mRNA | BC090903.1 | - | 5 - 6 | 977 | 61 | |
| BC165573.1 | - | 5 - 6 | 878 | 61 | ||
More Information
Set Membership: |
|

enpp6
Genomic Map