Assay Details
| Gene Symbol: sh3glb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017617.1 | NP_001017617.1 | 7 - 8 | 924 | 72 | |
| XM_009297515.3 | XP_009295790.1 | 9 - 10 | 1118 | 72 | ||
| GenBank mRNA | BC092831.1 | - | 7 - 8 | 924 | 72 | |
| BC164182.1 | - | 7 - 8 | 784 | 72 | ||
| CV488337.1 | - | 3 - 4 | 248 | 72 | ||
More Information
Set Membership: |
|

sh3glb1a
Genomic Map