Assay Details
| Gene Symbol: dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328370.1 | NP_001315299.1 | 5 - 6 | 485 | 78 | |
| XM_017357966.2 | XP_017213455.1 | 5 - 6 | 535 | 78 | ||
| GenBank mRNA | BC093237.1 | - | 5 - 6 | 467 | 78 | |
More Information
Set Membership: |
|

dusp11
Genomic Map