Assay Details
| Gene Symbol: c1qbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017858.2 | NP_001017858.2 | 2 - 3 | 477 | 80 | |
| GenBank mRNA | BC092751.1 | - | 2 - 3 | 424 | 80 | |
| BC164455.1 | - | 2 - 3 | 391 | 80 | ||
| EH440105.1 | - | 2 - 3 | 450 | 80 | ||
More Information
Set Membership: |
|

c1qbp
Genomic Map