Assay Details
| Gene Symbol: capn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018227.1 | NP_001018177.1 | 17 - 18 | 1879 | 81 | |
| GenBank mRNA | BC076522.1 | - | 18 - 19 | 1853 | 81 | |
| BC095045.1 | - | 17 - 18 | 1879 | 81 | ||
| BC155147.1 | - | 17 - 18 | 1830 | 81 | ||
| BC165647.1 | - | 17 - 18 | 1841 | 81 | ||
| BC171423.1 | - | 17 - 18 | 1830 | 81 | ||
More Information
Set Membership: |
|

capn2b
Genomic Map