Assay Details
| Gene Symbol: capn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018227.1 | NP_001018177.1 | 19 - 20 | 2082 | 73 | |
| GenBank mRNA | BC076522.1 | - | 20 - 21 | 2056 | 73 | |
| BC095045.1 | - | 19 - 20 | 2082 | 73 | ||
| BC155147.1 | - | 19 - 20 | 2033 | 73 | ||
| BC165647.1 | - | 19 - 20 | 2044 | 73 | ||
| BC171423.1 | - | 19 - 20 | 2033 | 73 | ||
More Information
Set Membership: |
|

capn2b
Genomic Map