Assay Details
| Gene Symbol: cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020500.1 | NP_001018336.1 | 2 - 3 | 438 | 116 | |
| GenBank mRNA | BC095170.1 | - | 2 - 3 | 438 | 116 | |
| BC153653.1 | - | 2 - 3 | 438 | 116 | ||
| BC165119.1 | - | 2 - 3 | 426 | 116 | ||
More Information
Set Membership: |
|

cybrd1
Genomic Map