Assay Details
| Gene Symbol: homer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020634.1 | NP_001018470.1 | 858 | 154 | ||
| GenBank mRNA | BC095619.1 | - | 858 | 154 | ||
| BC159234.1 | - | 923 | 154 | |||
More Information
Set Membership: |
|

homer2
Genomic Map