Assay Details
| Gene Symbol: prdx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024406.1 | NP_001019577.1 | 5 - 6 | 401 | 69 | |
| GenBank mRNA | BC095755.1 | - | 5 - 6 | 401 | 69 | |
More Information
Set Membership: |
|

prdx5
Genomic Map