Assay Details
| Gene Symbol: slc5a6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025528.2 | NP_001020699.1 | 3 - 4 | 631 | 74 | |
| XM_005158575.4 | XP_005158632.1 | 3 - 4 | 838 | 74 | ||
| XM_005158576.4 | XP_005158633.1 | 3 - 4 | 838 | 74 | ||
| GenBank mRNA | BC097129.1 | - | 3 - 4 | 632 | 74 | |
| BC155140.1 | - | 3 - 4 | 637 | 74 | ||
| BC171549.1 | - | 2 - 3 | 459 | 74 | ||
More Information
Set Membership: |
|

slc5a6b
Genomic Map