Assay Details
| Gene Symbol: ppp1r14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029963.1 | NP_001025134.1 | 1 - 2 | 305 | 63 | |
| GenBank mRNA | BC098532.1 | - | 1 - 2 | 305 | 63 | |
| BC159209.1 | - | 1 - 2 | 278 | 63 | ||
More Information
Set Membership: |
|

ppp1r14c
Genomic Map