Assay Details
| Gene Symbol: rgs7bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030069.1 | NP_001025240.1 | 1 - 2 | 355 | 146 | |
| GenBank mRNA | BC096790.1 | - | 1 - 2 | 355 | 146 | |
| BC152595.1 | - | 1 - 2 | 333 | 146 | ||
| BC165928.1 | - | 1 - 2 | 186 | 146 | ||
More Information
Set Membership: |
|

rgs7bpb
Genomic Map