Assay Details
| Gene Symbol: camk2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030125.1 | NP_001025296.1 | 17 - 18 | 1359 | 82 | |
| XM_005156365.4 | XP_005156422.1 | 18 - 19 | 1468 | 82 | ||
| XM_021480165.1 | XP_021335840.1 | 18 - 19 | 1458 | 82 | ||
| GenBank mRNA | BC096785.1 | - | 17 - 18 | 1359 | 82 | |
More Information
Set Membership: |
|

camk2g1
Genomic Map