Assay Details
| Gene Symbol: tdo2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102616.2 | NP_001096086.1 | 6 - 7 | 643 | 54 | |
| GenBank mRNA | BC150376.1 | - | 6 - 7 | 645 | 54 | |
| BC151920.1 | - | 6 - 7 | 645 | 54 | ||
More Information
Set Membership: |
|

tdo2a
Genomic Map