Assay Details
| Gene Symbol: gpank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105703.2 | NP_001099173.2 | 3 - 4 | 605 | 56 | |
| NM_001327760.1 | NP_001314689.1 | 3 - 4 | 589 | 56 | ||
| NM_001327761.1 | NP_001314690.1 | 3 - 4 | 600 | 56 | ||
| GenBank mRNA | BC151976.1 | - | 3 - 4 | 606 | 56 | |
More Information
Set Membership: |
|

gpank1
Genomic Map