Assay Details
| Gene Symbol: dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109694.2 | NP_001103164.1 | 9 - 10 | 1480 | 67 | |
| GenBank mRNA | BC162442.1 | - | 9 - 10 | 1480 | 67 | |
| BC163055.1 | - | 9 - 10 | 1480 | 67 | ||
| EU344799.1 | - | 9 - 10 | 1420 | 67 | ||
More Information
Set Membership: |
|

dbh
Genomic Map