Assay Details
| Gene Symbol: fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001812.1 | NP_001001812.1 | 6 - 7 | 609 | 63 | |
| GenBank mRNA | AY278107.1 | - | 6 - 7 | 613 | 63 | |
| AY424301.1 | - | 6 - 7 | 609 | 63 | ||
More Information
Set Membership: |
|

fshr
Genomic Map