Assay Details
| Gene Symbol: mrpl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002052.2 | NP_001002052.1 | 3 - 4 | 294 | 76 | |
| GenBank mRNA | BC071325.1 | - | 3 - 4 | 295 | 76 | |
| BC153659.1 | - | 3 - 4 | 326 | 76 | ||
More Information
Set Membership: |
|

mrpl23
Genomic Map