Assay Details
| Gene Symbol: mrpl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002052.2 | NP_001002052.1 | 4 - 5 | 364 | 79 | |
| GenBank mRNA | BC071325.1 | - | 4 - 5 | 365 | 79 | |
| BC153659.1 | - | 4 - 5 | 396 | 79 | ||
More Information
Set Membership: |
|

mrpl23
Genomic Map