Assay Details
| Gene Symbol: rpl5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002106.1 | NP_001002106.1 | 4 - 5 | 406 | 88 | |
| GenBank mRNA | BC071498.1 | - | 4 - 5 | 406 | 88 | |
| BC076208.1 | - | 4 - 5 | 406 | 88 | ||
More Information
Set Membership: |
|

rpl5b
Genomic Map