Assay Details
| Gene Symbol: nudt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002107.2 | NP_001002107.2 | 2 - 3 | 254 | 91 | |
| NM_001346166.1 | NP_001333095.1 | 2 - 3 | 410 | 91 | ||
| XM_009302185.3 | XP_009300460.1 | 2 - 3 | 374 | 91 | ||
| XM_017356464.2 | XP_017211953.1 | 2 - 3 | 307 | 91 | ||
| GenBank mRNA | BC071496.1 | - | 2 - 3 | 254 | 91 | |
More Information
Set Membership: |
|

nudt15
Genomic Map