Assay Details
| Gene Symbol: dcun1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002156.2 | NP_001002156.2 | 4 - 5 | 545 | 84 | |
| GenBank mRNA | BC071344.1 | - | 4 - 5 | 538 | 84 | |
More Information
Set Membership: |
|

dcun1d2b
Genomic Map